3,5-Dimethoxyzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3,5-Dimethoxyphenyl)propensäure, 3,5-Dimethoxycinnamsäure
OR11319-25G
APOSOR11319-25GEA
97.3
EUR
APOSOR11319-25G
3,5-Dimethoxyzimtsäure 98%
3,5-Dimethoxyzimtsäure
Formel:
C₁₁H₁₂O₄ Molecular Weight: 208.21 g/mol Siedepunkt: 343 Schmelzpunkt: 174…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004389 CAS-Nummer: 16909-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...