4-Amino-6-hydroxypyrimidine
Lieferant: Apollo Scientific
OR11321-5G
APOSOR11321-5GEA
55.1
EUR
APOSOR11321-5G
4-Amino-6-hydroxypyrimidine
4-Amino-6-hydroxypyrimidine
Formel:
C₄H₅N₃O Molecular Weight: 111,10 g/mol Schmelzpunkt: 262…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051502 CAS-Nummer: 1193-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...