3-(3,5-Dimethoxyphenyl)propionsäure

Lieferant: Apollo Scientific
Achtung

OR11325-100MG
APOSOR11325-100MGEA 21.9 EUR
APOSOR11325-100MG
3-(3,5-Dimethoxyphenyl)propionsäure
3-(3,5-Dimethoxyphenyl)propionsäure
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425685
CAS-Nummer: 717-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR