3-Methoxy-4-methylbenzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR11329-5G
OR11329-10G
OR11329-1G
APOSOR11329-5GEA
34.8
EUR
APOSOR11329-5G
APOSOR11329-10G
APOSOR11329-1G
3-Methoxy-4-methylbenzaldehyde
3-Methoxy-4-methylbenzaldehyde
Formel:
C9H10O2 Molecular Weight: 150.1700 g/mol Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114050 CAS-Nummer: 24973-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...