[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride
OR11342-1G
APOSOR11342-1GEA
88.6
EUR
APOSOR11342-1G
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride
Formel:
C₁₂H₈Cl₂O₂S Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631918 CAS-Nummer: 20443-74-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...