4-(2-Methoxyphenoxy)benzenesulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR11345-250MG
APOSOR11345-250MGEA 115 EUR
APOSOR11345-250MG
4-(2-Methoxyphenoxy)benzenesulphonyl chloride 95%
4-(2-Methoxyphenoxy)benzenesulphonyl chloride
Formel: C₁₃H₁₁ClO₄S
Molecular Weight: 298,75 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01631898
CAS-Nummer: 690632-30-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR