4-(3,4-Dichlorophenoxy)aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3',4'-dichlorodiphenyl ether

OR11352-5G
APOSOR11352-5GEA 169 EUR
APOSOR11352-5G
4-(3,4-Dichlorophenoxy)aniline
4-(3,4-Dichlorophenoxy)aniline
Formel: C₁₂H₉Cl₂NO
Molecular Weight: 254,112 g/mol
Schmelzpunkt: 68,7…70.3 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00459613
CAS-Nummer: 67651-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR