5-Chlor-2-pyridincarbonitril 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-cyanopyridine

OR11381-5G
APOSOR11381-5GEA 21.9 EUR
APOSOR11381-5G
5-Chlor-2-pyridincarbonitril 96%
5-Chloropicolinonitrile
Formel: C6H3ClN2
Molecular Weight: 138,55 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788835
CAS-Nummer: 89809-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR