2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one
OR11386-1G
APOSOR11386-1GEA
49.3
EUR
APOSOR11386-1G
2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one
2-Amino-5,5-dimethyl-5,6-dihydro-4H-benzothiazol-7-one
Formel:
C₉H₁₂N₂OS Molecular Weight: 196,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00170187 CAS-Nummer: 1629-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...