Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

OR11400-1G
APOSOR11400-1GEA 163 EUR
APOSOR11400-1G
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat 98%
Ethyl-8-brom-5-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₁BrN₂O₂
Molecular Weight: 283.1212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09475853
CAS-Nummer: 135995-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR