2-(Piperidin-1-ylsulfonyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Piperidin-1-ylsulphonyl)phenyl)boronic acid

OR11411-1G
APOSOR11411-1GEA 94.4 EUR
APOSOR11411-1G
2-(Piperidin-1-ylsulfonyl)phenylboronsäure 97%
2-(Piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₄S
Molecular Weight: 269,1300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09475844
CAS-Nummer: 957034-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR