1-Boc-7-bromindol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1H-indole, N-BOC protected

OR11414-1G
APOSOR11414-1GEA 125 EUR
APOSOR11414-1G
1-Boc-7-bromindol 98%
1-Boc-7-bromindol
Formel: C₁₃H₁₄BrNO₂
Molecular Weight: 296,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09475841
CAS-Nummer: 868561-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR