1,3-Dibrom-5-nitrobenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR11436-1G
OR11436-5G
OR11436-25G
APOSOR11436-1GEA
29
EUR
APOSOR11436-1G
APOSOR11436-5G
APOSOR11436-25G
1,3-Dibrom-5-nitrobenzol 97%
1,3-Dibromo-5-nitrobenzene
Formel:
C₆H₃Br₂NO₂ Schmelzpunkt: 100…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092529 CAS-Nummer: 6311-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...