4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Morpholinoethoxy)phenylboronic acid, pinacol, 4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)morpholine, 4-[2-(Morpholin-4-yl)ethoxy]benzeneboronic acid, pinacol ester

OR11446-250MG
APOSOR11446-250MGEA 29 EUR
APOSOR11446-250MG
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine
Formel: C₁₈H₂₈BNO₄
Molecular Weight: 333,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04440857
CAS-Nummer: 690636-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR