(Tetrahydropyran-4-yl)methyltosylat 98%

Lieferant: Apollo Scientific

Synonyme: (Tetrahydro-2H-pyran-4-yl)methyl 4-methylbenzenesulphonate

OR11456-1G
APOSOR11456-1GEA 39.3 EUR
APOSOR11456-1G
(Tetrahydropyran-4-yl)methyltosylat 98%
(Tetrahydropyran-4-yl)methyltosylat
Formel: C₁₃H₁₈O₄S
Molecular Weight: 270,3500 g/mol
MDL: MFCD09475806
CAS-Nummer: 101691-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR