tert-Butyl 4-bromobutanoate 95%

Lieferant: Apollo Scientific
Achtung

OR11460-1G OR11460-25G OR11460-5G
APOSOR11460-1GEA 21.9 EUR
APOSOR11460-1G APOSOR11460-25G APOSOR11460-5G
tert-Butyl 4-bromobutanoate 95%
tert-Butyl 4-bromobutanoate
Formel: C₈H₁₅BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02179416
CAS-Nummer: 110661-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR