3,5-Dicarboxyphenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dicarboxybenzeneboronic acid, 5-Boronoisophthalic acid

OR11481-25G OR11481-5G OR11481-100G
APOSOR11481-25GEA 174 EUR
APOSOR11481-25G APOSOR11481-5G APOSOR11481-100G
3,5-Dicarboxyphenylboronic acid 98%
3,5-Dicarboxyphenylboronic acid
Formel: C₈H₇BO₆
Molecular Weight: 209.95 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06203347
CAS-Nummer: 881302-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR