Ethyl-2-chlor-5-oxazolcarboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-chlorooxazole-5-carboxylate
OR11484-1G
APOSOR11484-1GEA
56.7
EUR
APOSOR11484-1G
Ethyl-2-chlor-5-oxazolcarboxylat
Ethyl-2-chlor-5-oxazolcarboxylat
Formel:
C₆H₆ClNO₃ Molecular Weight: 175,5697 g/mol |
MDL:
MFCD09475866 CAS-Nummer: 862599-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...