4-Brom-2-thiophencarbonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromothiophene-2-carbonitrile, 4-Bromo-2-cyanothiophene

OR11486-1G
APOSOR11486-1GEA 21.9 EUR
APOSOR11486-1G
4-Brom-2-thiophencarbonitril 98%
4-Brom-2-thiophencarbonitril
Formel: C₅H₂BrNS
Molecular Weight: 188,0500 g/mol
Schmelzpunkt: 49 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00114951
CAS-Nummer: 18791-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR