3-Methyl-2-thiophencarbonitril 95%

Lieferant: Apollo Scientific

OR11488-25G OR11488-5G
APOSOR11488-25GEA 237 EUR
APOSOR11488-25G APOSOR11488-5G
3-Methyl-2-thiophencarbonitril 95%
3-Methyl-2-thiophencarbonitril
Formel: C₆H₅NS
Siedepunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052086
CAS-Nummer: 55406-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR