2-Brom-4'-(1-pyrrolidinyl)acetophenon 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Pyrrolidin-1-yl)phenacyl bromide
OR11500-1G
OR11500-5G
OR11500-25G
APOSOR11500-1GEA
79.9
EUR
APOSOR11500-1G
APOSOR11500-5G
APOSOR11500-25G
2-Brom-4'-(1-pyrrolidinyl)acetophenon 97%
2-Brom-4'-(1-pyrrolidinyl)acetophenon
Formel:
C₁₂H₁₄BrNO Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075690 CAS-Nummer: 216144-18-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...