3-(Hydroxymethyl)phenylboronsäurepinakolester 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Hydroxymethyl)benzolboronsäure(tetramethylethylen)ester

OR11503-25G
APOSOR11503-25GEA 97.3 EUR
APOSOR11503-25G
3-(Hydroxymethyl)phenylboronsäurepinakolester 96%
3-(Hydroxymethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₃
Molecular Weight: 234.10 g/mol
Schmelzpunkt: 95…99 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09266196
CAS-Nummer: 443776-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR