AMPA

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Hydroxy-5-methylisoxazol-4-yl)-DL-alanine

OR1150T-10MG
APOSOR1150T-10MGEA 232 EUR
APOSOR1150T-10MG
AMPA
AMPA

A selective agonist of the exitatory neurotransmitter L-glutamic acid. AMPA does not interfere with binding sites for kainic acid in vitro.

Formel: C₇H₁₀N₂O₄
Molecular Weight: 186,2 g/mol
Schmelzpunkt: >200 °C (Zersetzung)
Lagertemperatur: Gefrierschrank
CAS-Nummer: 77521-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR