2-Chloro-6-(1H-pyrazol-1-yl)pyrazine 95%

Lieferant: Apollo Scientific
Achtung

OR11510-5G
APOSOR11510-5GEA 49.3 EUR
APOSOR11510-5G
2-Chloro-6-(1H-pyrazol-1-yl)pyrazine 95%
2-Chloro-6-(1H-pyrazol-1-yl)pyrazine
Formel: C₇H₅ClN₄
Molecular Weight: 180.594 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800958
CAS-Nummer: 642459-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR