3-Brom-2-chlortoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR11515-1G
APOSOR11515-1GEA
21.9
EUR
APOSOR11515-1G
3-Brom-2-chlortoluol 98%
3-Brom-2-chlortoluol
Formel:
C₇H₆BrCl Molecular Weight: 205,48 g/mol Siedepunkt: 125…135 °C/ 50mmHg Schmelzpunkt: 18 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782165 CAS-Nummer: 97329-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...