(tert-Butyl) 6-bromonicotinate 98%

Lieferant: Apollo Scientific
Achtung

OR11518-1G OR11518-500MG
APOSOR11518-1GEA 174 EUR
APOSOR11518-1G APOSOR11518-500MG
(tert-Butyl) 6-bromonicotinate 98%
(tert-Butyl) 6-bromonicotinate
Formel: C₁₀H₁₂BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09264580
CAS-Nummer: 941294-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR