5-Brom-2-iod-4-methylpyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Brom-2-iod-4-picolin

OR11521-1G
APOSOR11521-1GEA 177 EUR
APOSOR11521-1G
5-Brom-2-iod-4-methylpyridin 98%
5-Brom-2-iod-4-methylpyridin
Formel: C₆H₅BrIN
Molecular Weight: 297,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801049
CAS-Nummer: 941294-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR