2-Chlor-6-nitroanisol 98%

Lieferant: Apollo Scientific
Achtung

OR11522-25G OR11522-100G
APOSOR11522-25GEA 59.6 EUR
APOSOR11522-25G APOSOR11522-100G
2-Chlor-6-nitroanisol 98%
2-Chlor-6-nitroanisol
Formel: C₇H₆ClNO₃
Molecular Weight: 187.58 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065066
CAS-Nummer: 80866-77-9
EINECS: 279-580-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR