Imidazo[1,2-a]pyridin-8-ol Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

OR11533-1G
APOSOR11533-1GEA 58 EUR
APOSOR11533-1G
Imidazo[1,2-a]pyridin-8-ol Hydrochlorid 97%
Imidazo[1,2-a]pyridin-8-ol Hydrochlorid
Formel: C₇H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09800942
CAS-Nummer: 100592-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR