2-Chloro-4-(pyrrolidin-1-yl)pyrimidine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR11552-250MG
APOSOR11552-250MGEA
0
EUR
APOSOR11552-250MG
2-Chloro-4-(pyrrolidin-1-yl)pyrimidine 98%
2-Chloro-4-(pyrrolidin-1-yl)pyrimidine
Formel:
C₈H₁₀ClN₃ Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475891 CAS-Nummer: 35691-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...