5-Methyl-2-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methoxy-3-methyl-6-nitrobenzol, 3-Methoxy-4-nitrotoluol, Methyl-6-nitro-m-tolylether
OR11561-5G
APOSOR11561-5GEA
21.9
EUR
APOSOR11561-5G
5-Methyl-2-nitroanisol 98%
5-Methyl-2-nitroanisol
Formel:
C₈H₉NO₃ Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007367 CAS-Nummer: 38512-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...