4-Brom-N-tert-butylbenzamid 98%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-N-(tert-butyl)benzamide

OR11562-5G
APOSOR11562-5GEA 77 EUR
APOSOR11562-5G
4-Brom-N-tert-butylbenzamid 98%
4-Brom-N-tert-butylbenzamid
Formel: C₁₁H₁₄BrNO
Molecular Weight: 256,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00462539
CAS-Nummer: 42498-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR