3-Brom-N-methylbenzamid 98%

Lieferant: Apollo Scientific
Achtung

OR11565-250MG
APOSOR11565-250MGEA 21.9 EUR
APOSOR11565-250MG
3-Brom-N-methylbenzamid 98%
3-Brom-N-methylbenzamid
Formel: C₈H₈BrNO
Molecular Weight: 214.0592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01060879
CAS-Nummer: 49834-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR