4-[N-(3-Methylbutanoyl)sulphamoyl]benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(N-(3-Methylbutanoyl)sulphamoyl)phenyl)boronic acid

OR11566-250MG
APOSOR11566-250MGEA 46.4 EUR
APOSOR11566-250MG
4-[N-(3-Methylbutanoyl)sulphamoyl]benzeneboronic acid 98%
4-[N-(3-Methylbutanoyl)sulphamoyl]benzeneboronic acid
Formel: C₁₁H₁₆BNO₅S
Molecular Weight: 285,124 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800865
CAS-Nummer: 957120-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR