5-Brom-4-ethylpyrimidin

Lieferant: Apollo Scientific
Achtung

OR11580-1G
APOSOR11580-1GEA 40.6 EUR
APOSOR11580-1G
5-Brom-4-ethylpyrimidin
5-Bromo-4-ethylpyrimidine
Formel: C₆H₇BrN₂
Molecular Weight: 187,0372 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800935
CAS-Nummer: 951884-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR