5-Brom-4-pentylpyrimidin

Lieferant: Apollo Scientific
Achtung

OR11586-250MG
APOSOR11586-250MGEA 29 EUR
APOSOR11586-250MG
5-Brom-4-pentylpyrimidin
5-Bromo-4-pentylpyrimidine
Formel: C₉H₁₃BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800934
CAS-Nummer: 951884-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR