Methyl-5-bromo-1H-pyrrole-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-bromo-1H-pyrrole-2-carboxylate
OR11588-1G
APOSOR11588-1GEA
62.5
EUR
APOSOR11588-1G
Methyl-5-bromo-1H-pyrrole-2-carboxylate
Methyl-5-bromo-1H-pyrrole-2-carboxylate
Formel:
C₆H₆BrNO₂ Molecular Weight: 204.0213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801030 CAS-Nummer: 934-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...