2-Bromtoluol-4-sulfonamid 98%

Lieferant: Apollo Scientific
Achtung

OR11601-1G
APOSOR11601-1GEA 130 EUR
APOSOR11601-1G
2-Bromtoluol-4-sulfonamid 98%
2-Bromtoluol-4-sulfonamid
Formel: C₇H₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09801033
CAS-Nummer: 210824-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR