6-Chloro-N-cyclohexyl-4-pyrimidinamine 98%

Lieferant: Apollo Scientific
Achtung

OR11612-250MG
APOSOR11612-250MGEA 0 EUR
APOSOR11612-250MG
6-Chloro-N-cyclohexyl-4-pyrimidinamine 98%
6-Chloro-N-cyclohexyl-4-pyrimidinamine
Formel: C₁₀H₁₄ClN₃
Lagertemperatur: Kühlschrank
MDL: MFCD03669655
CAS-Nummer: 61667-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR