3-Chlor-6-(methylamino)pyridazin
Lieferant: Apollo Scientific
|
Achtung
|
OR11643-250MG
APOSOR11643-250MGEA
21.9
EUR
APOSOR11643-250MG
3-Chlor-6-(methylamino)pyridazin
3-Chloro-6-(methylamino)pyridazine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,5700 g/mol Schmelzpunkt: 195…201 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02178601 CAS-Nummer: 14959-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...