Methyl-6-amino-3-brompicolinat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-amino-3-bromopicolinate, Methyl 6-amino-3-bromopyridine-2-carboxylate

OR11647-5G
APOSOR11647-5GEA 78.3 EUR
APOSOR11647-5G
Methyl-6-amino-3-brompicolinat
Methyl-6-amino-3-brompicolinat
Formel: C₇H₇BrN₂O₂
Schmelzpunkt: 95…97 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09800897
CAS-Nummer: 178876-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR