3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR11656-100MG
APOSOR11656-100MGEA 21.9 EUR
APOSOR11656-100MG
3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure
3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₄
Schmelzpunkt: 143…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09800860
CAS-Nummer: 957120-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR