3-Brom-5-nitrobenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR11657-1G
APOSOR11657-1GEA 34.8 EUR
APOSOR11657-1G
3-Brom-5-nitrobenzonitril 98%
3-Brom-5-nitrobenzonitril
Formel: C₇H₃BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00463804
CAS-Nummer: 49674-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR