Chlorogenic acid 98+%

Lieferant: Apollo Scientific

Synonyme: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

OR1166-25G OR1166-1G OR1166-5G
APOSOR1166-25GEA 155 EUR
APOSOR1166-25G APOSOR1166-1G APOSOR1166-5G
Chlorogenic acid 98+%
Chlorogenic acid
Formel: C₁₆H₁₈O₉
Molecular Weight: 354.31 g/mol
Schmelzpunkt: 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003862
CAS-Nummer: 327-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR