3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chloro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 3-Chloro-5-(piperidin-1-carbonyl)phenylboronic acid

OR11661-250MG
APOSOR11661-250MGEA 62.5 EUR
APOSOR11661-250MG
3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure
3-Chlor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
Molecular Weight: 267,5200 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09800859
CAS-Nummer: 957120-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR