8-Brom-6-methylimidazo[1,2-a]pyridin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR11663-250MG
APOSOR11663-250MGEA 29 EUR
APOSOR11663-250MG
8-Brom-6-methylimidazo[1,2-a]pyridin Hydrochlorid
8-Brom-6-methylimidazo[1,2-a]pyridin Hydrochlorid
Formel: C₈H₈BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800972
CAS-Nummer: 957120-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR