6-Bromo-2-chloro-3-methoxybenzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-2-chloro-3-methoxyphenylboronic acid, (6-Bromo-2-chloro-3-methoxyphenyl)boronic acid

OR11674-1G
APOSOR11674-1GEA 68.3 EUR
APOSOR11674-1G
6-Bromo-2-chloro-3-methoxybenzeneboronic acid
6-Bromo-2-chloro-3-methoxybenzeneboronic acid
Formel: C₇H₇BBrClO₃
Molecular Weight: 265,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800872
CAS-Nummer: 957062-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR