Ethyl-8-brom-4-hydroxychinolin-3-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 8-bromo-4-hydroxyquinoline-3-carboxylate

OR11682-1G
APOSOR11682-1GEA 21.9 EUR
APOSOR11682-1G
Ethyl-8-brom-4-hydroxychinolin-3-carboxylat
Ethyl-8-brom-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀BrNO₃
Molecular Weight: 296,1167 g/mol
Schmelzpunkt: 256…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173380
CAS-Nummer: 35975-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR