5-Brom-2-methoxy-3-nitropyridin 97%

Lieferant: Apollo Scientific
Achtung

OR11688-1G
APOSOR11688-1GEA 21.9 EUR
APOSOR11688-1G
5-Brom-2-methoxy-3-nitropyridin 97%
5-Brom-2-methoxy-3-nitropyridin
Formel: C₆H₅BrN₂O₃
Molecular Weight: 233,0200 g/mol
Schmelzpunkt: 89…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07374968
CAS-Nummer: 152684-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR