5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine

Lieferant: Apollo Scientific
Achtung

OR11690-250MG
APOSOR11690-250MGEA 21.9 EUR
APOSOR11690-250MG
5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine
5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine
Formel: C₁₀H₁₁BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09801066
CAS-Nummer: 742085-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR